siRNA Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145456.2 | NP_663431.2 | 14 | 3476 | ||
| GenBank mRNA | AK037945.1 | - | 10 | 2071 | ||
| BC021311.1 | - | 5 | 1153 | |||
| BC021521.1 | - | 5 | 1356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim6| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145456.2 | NP_663431.2 | 14 | 3476 | ||
| GenBank mRNA | AK037945.1 | - | 10 | 2071 | ||
| BC021311.1 | - | 5 | 1153 | |||
| BC021521.1 | - | 5 | 1356 | |||