siRNA Details
| Gene Symbol: Uqcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026063.2 | NP_080339.1 | 3 | 267 | ||
| GenBank mRNA | AK013521.1 | - | 3 | 271 | ||
| AK019014.1 | - | 3 | 268 | |||
| BC030629.1 | - | 3 | 270 | 1054665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcc2| Gene Symbol: Uqcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026063.2 | NP_080339.1 | 3 | 267 | ||
| GenBank mRNA | AK013521.1 | - | 3 | 271 | ||
| AK019014.1 | - | 3 | 268 | |||
| BC030629.1 | - | 3 | 270 | 1054665 | ||