siRNA Details
| Gene Symbol: Cmtm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026066.2 | NP_080342.1 | 4 | 585 | ||
| NM_029390.1 | NP_083666.1 | 3 | 405 | |||
| GenBank mRNA | AK005168.1 | - | 3 | 448 | ||
| AK013666.1 | - | 4 | 585 | |||
| AY241867.1 | - | 4 | 575 | |||
| BC049665.1 | - | 4 | 572 | 6533928 | ||
| DQ115983.1 | - | 3 | 405 | |||

Cmtm5
Genomic Map