siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024197.1 | NP_077159.1 | 3 | 294 | ||
| GenBank mRNA | AK005339.1 | - | 3 | 330 | ||
| AK013675.1 | - | 3 | 338 | |||
| AK046292.1 | - | 198 | ||||
| AK145979.1 | - | 3 | 350 | |||
| AK177419.1 | - | 2 | 148 | |||
| AK184477.1 | - | 2 | 148 | |||
| BC003439.1 | - | 3 | 294 | 2647815 | ||

Ndufa10
Genomic Map