siRNA Details
| Gene Symbol: Med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026068.2 | NP_080344.2 | 4 | 893 | ||
| GenBank mRNA | AK005655.1 | - | 4 | 302 | ||
| AK013990.1 | - | 4 | 334 | |||
| BC051929.1 | - | 4 | 893 | 5709831 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med31| Gene Symbol: Med31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026068.2 | NP_080344.2 | 4 | 893 | ||
| GenBank mRNA | AK005655.1 | - | 4 | 302 | ||
| AK013990.1 | - | 4 | 334 | |||
| BC051929.1 | - | 4 | 893 | 5709831 | ||