siRNA Details
| Gene Symbol: Ift22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026073.3 | NP_080349.1 | 2 | 177 | ||
| GenBank mRNA | AK005820.1 | - | 2 | 137 | ||
| AK160464.1 | - | 2 | 188 | |||
| BC009150.1 | - | 2 | 177 | 2651447 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift22| Gene Symbol: Ift22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026073.3 | NP_080349.1 | 2 | 177 | ||
| GenBank mRNA | AK005820.1 | - | 2 | 137 | ||
| AK160464.1 | - | 2 | 188 | |||
| BC009150.1 | - | 2 | 177 | 2651447 | ||