siRNA Details
| Gene Symbol: Rab3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023852.5 | NP_076341.1 | 3, 4 | 560 | ||
| GenBank mRNA | AB232589.1 | - | 3, 4 | 372 | ||
| AF312037.1 | - | 3, 4 | 372 | |||
| AJ310532.1 | - | 3, 4 | 372 | |||
| AK014050.1 | - | 3, 4 | 539 | |||
| AK014132.1 | - | 3, 4 | 482 | |||
| AK079574.1 | - | 3, 4 | 457 | |||
| AK148904.1 | - | 3, 4 | 481 | |||
| AK161006.1 | - | 3, 4 | 458 | |||
| AK164316.1 | - | 3, 4 | 505 | |||
| AY026947.1 | - | 3, 4 | 441 | |||
| BC114335.2 | - | 3, 4 | 433 | 40045744 | ||

Rab3c
Genomic Map