siRNA Details
| Gene Symbol: Socs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080843.2 | NP_543119.2 | 2 | 1179 | ||
| GenBank mRNA | AK014115.1 | - | 2 | 1209 | ||
| AY046322.1 | - | 1 | 1007 | |||
| BC117814.1 | - | 1 | 1100 | 40092681 | ||
| BC117815.1 | - | 1 | 1100 | 40092683 | ||

Socs4
Genomic Map