siRNA Details
| Gene Symbol: Gprasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004359.2 | NP_001004359.1 | 10 | 2324 | ||
| NM_001005385.1 | NP_001005385.1 | 10 | 2210 | |||
| NM_026081.5 | NP_080357.4 | 11 | 2413 | |||
| GenBank mRNA | AB093241.1 | - | 5 | 1530 | ||
| AK050919.1 | - | 7 | 1848 | |||
| AK083192.1 | - | 10 | 2170 | |||
| BC058970.1 | - | 1 | 1167 | 6826050 | ||
| BC070413.1 | - | 11 | 2361 | |||
| BC085157.1 | - | 10 | 2112 | 30357108 | ||

Gprasp1
Genomic Map