siRNA Details
Gene Symbol: Cltc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003908.1 | NP_001003908.1 | 13 | 2266 | ||
GenBank mRNA | AK122193.1 | - | 13 | 2270 | ||
AK143503.1 | - | 2 | 157 | |||
BC035544.1 | - | 8 | 1145 | |||
BC079897.1 | - | 13 | 2266 | 30546407 |
