siRNA Details
| Gene Symbol: Zc3h13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026083.2 | NP_080359.2 | 18 | 5305 | ||
| GenBank mRNA | AK016264.1 | - | 2 | 277 | ||
| BC023707.1 | - | 5 | 1173 | |||
| BC082576.1 | - | 7 | 2817 | |||
| BC094575.1 | - | 11 | 3854 | |||
| BC138264.1 | - | 18 | 5257 | 8861283 | ||
| BG062845.2 | - | 2 | 321 | |||

Zc3h13
Genomic Map