siRNA Details
| Gene Symbol: Zc3h13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026083.2 | NP_080359.2 | 11 | 2385 | ||
| GenBank mRNA | AK078225.1 | - | 11 | 2376 | ||
| BC038882.1 | - | 4 | 1195 | |||
| BC094575.1 | - | 3 | 989 | |||
| BC138264.1 | - | 11 | 2334 | 8861283 | ||

Zc3h13
Genomic Map