siRNA Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173181.3 | NP_775273.1 | 9 | 935 | ||
| GenBank mRNA | AK014202.1 | - | 9 | 870 | ||
| AK034783.1 | - | 2 | 81 | |||
| AK083941.1 | - | 9 | 879 | |||
| AK085187.1 | - | 9 | 873 | |||
| AK170609.1 | - | 8 | 768 | |||
| BC044809.1 | - | 9 | 857 | |||
| BC087731.1 | - | 899 | 6850296 | |||

Zc2hc1a
Genomic Map