siRNA Details
| Gene Symbol: Iqcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026090.2 | NP_080366.2 | 3 | 518 | ||
| GenBank mRNA | AK006688.1 | - | 3 | 517 | ||
| BC048652.1 | - | 3 | 523 | 6772797 | ||
| BC061014.1 | - | 3 | 518 | 6770556 | ||
| DQ153246.1 | - | 3 | 547 | |||

Iqcf4
Genomic Map