siRNA Details
| Gene Symbol: Iqcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026090.2 | NP_080366.2 | 3 | 669 | ||
| GenBank mRNA | BC048652.1 | - | 3 | 675 | 6772797 | |
| BC061014.1 | - | 3 | 669 | 6770556 | ||
| DQ153246.1 | - | 3 | 698 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcf4| Gene Symbol: Iqcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026090.2 | NP_080366.2 | 3 | 669 | ||
| GenBank mRNA | BC048652.1 | - | 3 | 675 | 6772797 | |
| BC061014.1 | - | 3 | 669 | 6770556 | ||
| DQ153246.1 | - | 3 | 698 | |||