siRNA Details
| Gene Symbol: 1700037H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026091.2 | NP_080367.1 | 4 | 427 | ||
| GenBank mRNA | AK003406.1 | - | 4 | 428 | ||
| AK006622.1 | - | 4 | 497 | |||
| AK040365.1 | - | 1 | 833 | |||
| BC006041.1 | - | 3 | 339 | 3497476 | ||
| BC071212.1 | - | 5 | 543 | |||

1700037H04Rik
Genomic Map