siRNA Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038635.2 | NP_001033724.2 | 2 | 884 | ||
| NM_183262.3 | NP_899085.3 | 2 | 884 | |||
| GenBank mRNA | AK006778.1 | - | 2 | 649 | ||
| AK165988.1 | - | 2 | 779 | |||
| BC047277.1 | - | 1 | 404 | |||
| BC138904.1 | - | 2 | 791 | 8861926 | ||
| BC138905.1 | - | 2 | 788 | 8861927 | ||

Stk35
Genomic Map