siRNA Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038635.2 | NP_001033724.2 | 3 | 1217 | ||
| NM_183262.3 | NP_899085.3 | 4 | 1967 | |||
| GenBank mRNA | AK165988.1 | - | 3 | 1112 | ||
| BC138904.1 | - | 4 | 1869 | 8861926 | ||
| BC138905.1 | - | 4 | 1866 | 8861927 | ||

Stk35
Genomic Map