siRNA Details
| Gene Symbol: Herc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026101.4 | NP_080377.2 | 5 | 907 | ||
| NM_030114.2 | NP_084390.1 | 5 | 907 | |||
| GenBank mRNA | AK009436.1 | - | 5 | 762 | ||
| AK019546.1 | - | 5 | 903 | |||
| AK087864.1 | - | 1 | 2977 | |||
| AK143111.1 | - | 5 | 781 | |||
| AK147723.1 | - | 5 | 794 | |||
| BC043082.1 | - | 5 | 719 | 6417111 | ||
| BC060033.1 | - | 3 | 492 | 6478123 | ||

Herc4
Genomic Map