siRNA Details
| Gene Symbol: Herc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026101.4 | NP_080377.2 | 19 | 2748 | ||
| NM_030114.2 | NP_084390.1 | 20 | 2772 | |||
| GenBank mRNA | AK019546.1 | - | 17 | 2381 | ||
| AK143111.1 | - | 20 | 2646 | |||
| AK145735.1 | - | 14 | 1696 | |||
| AK147723.1 | - | 19 | 2635 | |||
| BC026855.1 | - | 5 | 505 | |||
| BC043082.1 | - | 19 | 2560 | 6417111 | ||
| BC047157.1 | - | 5 | 650 | |||
| BC060033.1 | - | 18 | 2357 | 6478123 | ||

Herc4
Genomic Map