siRNA Details
| Gene Symbol: Tmco5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026104.4 | NP_080380.1 | 10, 11 | 751 | ||
| GenBank mRNA | AK007078.1 | - | 10, 11 | 735 | ||
| AK007251.1 | - | 9, 10 | 689 | |||
| BC049568.1 | - | 10, 11 | 715 | 6742928 | ||

Tmco5
Genomic Map