siRNA Details
| Gene Symbol: 1700093K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110133.1 | NP_001103603.1 | 5 | 1032 | ||
| NM_026105.3 | NP_080381.2 | 5 | 714 | |||
| GenBank mRNA | AK006386.1 | - | 2 | 218 | ||
| AK007139.1 | - | 4 | 471 | |||
| BC049567.1 | - | 5 | 714 | 6773879 | ||

1700093K21Rik
Genomic Map