siRNA Details
| Gene Symbol: Qpctl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026111.3 | NP_080387.2 | 3 | 414 | ||
| GenBank mRNA | AK007555.1 | - | 3 | 462 | ||
| AK007701.1 | - | 3 | 442 | |||
| AK035587.1 | - | 3 | 414 | |||
| AK045687.1 | - | 3 | 418 | |||
| AK148755.1 | - | 1 | 426 | |||
| BC058181.1 | - | 3 | 437 | 5325516 | ||

Qpctl
Genomic Map