siRNA Details
| Gene Symbol: 2210010C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023333.4 | NP_075822.3 | 3 | 299 | ||
| GenBank mRNA | AK007773.1 | - | 3 | 297 | ||
| AK008460.1 | - | 3 | 300 | |||
| AK008695.1 | - | 3 | 298 | |||
| BC061093.1 | - | 3 | 300 | 30306963 | ||

2210010C04Rik
Genomic Map