siRNA Details
| Gene Symbol: MAOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000898.4 | NP_000889.3 | 2, 3 | 309 | ||
| XM_005272608.3 | XP_005272665.1 | 4, 5 | 409 | |||
| XM_017029523.1 | XP_016885012.1 | 4, 5 | 459 | |||
| XM_017029524.1 | XP_016885013.1 | 3, 4 | 269 | |||
| GenBank mRNA | AK294306.1 | - | 3, 4 | 227 | ||
| AK298942.1 | - | 2, 3 | 246 | |||
| AK310458.1 | - | 2, 3 | 281 | |||
| AK312679.1 | - | 2, 3 | 284 | |||
| AU127841.1 | - | 2, 3 | 282 | |||
| BC022494.1 | - | 2, 3 | 255 | 4800208 | ||
| DC326844.1 | - | 2, 3 | 310 | |||
| M69177.1 | - | 2, 3 | 210 | |||
| S62734.1 | - | 2, 3 | 133 | |||

MAOB
Genomic Map