siRNA Details
| Gene Symbol: Unc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026123.3 | NP_080399.1 | 5 | 914 | ||
| GenBank mRNA | AK003273.1 | - | 5 | 689 | ||
| AK004433.1 | - | 5 | 678 | |||
| AK132757.1 | - | 5 | 915 | |||
| AK135885.1 | - | 5 | 906 | |||
| AK210936.1 | - | 2 | 92 | |||
| AK215289.1 | - | 1 | 17 | |||
| AY017214.1 | - | 5 | 899 | |||
| BC019484.1 | - | 5 | 881 | 4039903 | ||

Unc50
Genomic Map