siRNA Details
| Gene Symbol: 1110008F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026124.3 | NP_080400.1 | 1, 2 | 298 | ||
| GenBank mRNA | AK002452.1 | - | 1, 2 | 256 | ||
| AK003568.1 | - | 1, 2 | 293 | |||
| AK031002.1 | - | 1, 2 | 276 | |||
| AK179243.1 | - | 1, 2 | 19 | |||
| AK186989.1 | - | 1, 2 | 19 | |||
| BC019445.1 | - | 1, 2 | 236 | 5125733 | ||

1110008F13Rik
Genomic Map