siRNA Details
| Gene Symbol: Fam132a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026125.3 | NP_080401.1 | 4 | 688 | ||
| GenBank mRNA | AK002876.1 | - | 4 | 670 | ||
| AK003510.1 | - | 4 | 448 | |||
| AK004116.1 | - | 4 | 683 | |||
| AK007335.1 | - | 4 | 673 | |||
| BC026939.1 | - | 4 | 660 | 3966551 | ||
| DQ002404.1 | - | 4 | 421 | |||

Fam132a
Genomic Map