siRNA Details
| Gene Symbol: 4833420G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113550.1 | NP_001107022.1 | 2 | 619 | ||
| NM_026127.4 | NP_080403.4 | 3 | 682 | |||
| GenBank mRNA | AK008565.1 | - | 2 | 620 | ||
| AK008588.1 | - | 2 | 618 | |||
| AK014735.1 | - | 2 | 542 | |||
| AK146515.1 | - | 2 | 610 | |||
| AK149119.1 | - | 3 | 1472 | |||
| AK160863.1 | - | 2 | 603 | |||
| BC021327.1 | - | 3 | 662 | 5066171 | ||
| BC028328.1 | - | 3 | 1451 | |||
| BC030738.1 | - | 3 | 650 | |||
 Back To Top
 Back To Top
 4833420G17Rik
4833420G17Rik Genomic Map
Genomic Map Back To Top
 Back To Top