siRNA Details
| Gene Symbol: Adck3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163290.1 | NP_001156762.1 | 3 | 250 | ||
| NM_023341.3 | NP_075830.2 | 3 | 348 | |||
| GenBank mRNA | AJ278735.1 | - | 3 | 230 | ||
| AK004791.1 | - | 3 | 290 | |||
| AK014605.2 | - | 3 | 290 | |||
| AK179829.1 | - | 2 | 44 | |||
| AK179958.1 | - | 2 | 44 | |||
| BC030937.1 | - | 3 | 296 | 4922892 | ||
| BY261815.1 | - | 3 | 348 | |||
| BY275962.1 | - | 3 | 341 | |||

Adck3
Genomic Map