siRNA Details
| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026155.3 | NP_080431.1 | 4 | 399 | ||
| GenBank mRNA | AK002818.1 | - | 4 | 400 | ||
| AK029353.1 | - | 4 | 400 | |||
| AK076235.1 | - | 4 | 432 | |||
| AK088555.1 | - | 4 | 404 | |||
| AK150255.1 | - | 4 | 393 | |||
| AK162235.1 | - | 2 | 103 | |||
| BC011111.1 | - | 4 | 406 | 3965897 | ||
| BC020335.1 | - | 4 | 371 | |||
| CT010172.1 | - | 4 | 383 | |||

Ssr3
Genomic Map