siRNA Details
| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026157.2 | NP_080433.1 | 9 | 1593 | ||
| GenBank mRNA | AK002680.1 | - | 9 | 1564 | ||
| AK011546.1 | - | 9 | 1594 | |||
| AK036760.1 | - | 9 | 1598 | |||
| AK076565.1 | - | 9 | 1583 | |||
| AK135537.1 | - | 9 | 1561 | |||
| AK154747.1 | - | 9 | 1594 | |||
| AK166038.1 | - | 9 | 1571 | |||
| BC017535.1 | - | 9 | 1586 | |||
| BC057643.1 | - | 9 | 1567 | 2616306 | ||

Mtpap
Genomic Map