siRNA Details
| Gene Symbol: Plxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026162.6 | NP_080438.2 | 1, 2 | 932 | ||
| GenBank mRNA | AF378761.1 | - | 1, 2 | 131 | ||
| AK004640.1 | - | 1, 2 | 914 | |||
| AK019482.2 | - | 1, 2 | 888 | |||
| AK035165.1 | - | 1, 2 | 933 | |||
| AK154921.1 | - | 1, 2 | 879 | |||
| BC057881.1 | - | 1, 2 | 192 | 5349869 | ||

Plxdc2
Genomic Map