siRNA Details
| Gene Symbol: Pnpla8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026164.2 | NP_080440.2 | 4 | 1459 | ||
| GenBank mRNA | AB044139.1 | - | 4 | 1338 | ||
| AK004621.1 | - | 4 | 1426 | |||
| AK084928.1 | - | 1126 | ||||
| AK145776.1 | - | 4 | 1369 | |||
| AK163211.1 | - | 4 | 1427 | |||
| AK168475.1 | - | 4 | 1422 | |||
| BC127056.1 | - | 3 | 1319 | 40109753 | ||

Pnpla8
Genomic Map