siRNA Details
| Gene Symbol: Poc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026173.3 | NP_080449.1 | 4 | 570 | ||
| GenBank mRNA | AK004769.2 | - | 4 | 498 | ||
| AK012926.1 | - | 4 | 538 | |||
| AK089013.1 | - | 4 | 506 | |||
| AK098149.1 | - | 4 | 463 | |||
| AK150656.1 | - | 4 | 571 | |||
| BC016527.1 | - | 4 | 484 | 3157281 | ||
| BC027118.1 | - | 4 | 508 | 5359153 | ||
| BC037631.1 | - | 4 | 527 | 4954391 | ||

Poc5
Genomic Map