siRNA Details
| Gene Symbol: MARK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286124.1 | NP_001273053.1 | 12 | 1777 | ||
| NM_001286126.1 | NP_001273055.1 | 12 | 1777 | |||
| NM_001286128.1 | NP_001273057.1 | 11 | 1711 | |||
| NM_018650.4 | NP_061120.3 | 12 | 1777 | |||
| XM_005273134.4 | XP_005273191.1 | 12 | 1644 | |||
| XM_006711326.3 | XP_006711389.1 | 12 | 1038 | |||
| XM_011509561.2 | XP_011507863.1 | 12 | 1773 | |||
| XM_011509562.2 | XP_011507864.1 | 12 | 1390 | |||
| XM_017001305.1 | XP_016856794.1 | 13 | 1234 | |||
| XM_017001306.1 | XP_016856795.1 | 12 | 1274 | |||
| XM_017001307.1 | XP_016856796.1 | 13 | 1674 | |||
| GenBank mRNA | AB040910.1 | - | 11 | 1668 | ||
| AF154845.1 | - | 12 | 1403 | |||
| AK027493.1 | - | 8 | 734 | |||
| AK295506.1 | - | 12 | 1275 | |||
| AK314614.1 | - | 12 | 1437 | |||
| BC113869.1 | - | 12 | 1313 | 40036731 | ||
| BC114478.1 | - | 12 | 1313 | 40036730 | ||
| KJ534879.1 | - | 12 | 1137 | |||
| KJ534880.1 | - | 6 | 537 | |||
| KJ534881.1 | - | 12 | 1137 | |||
| KJ535089.1 | - | 12 | 1136 | |||

MARK1
Genomic Map