siRNA Details
| Gene Symbol: Gpalpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026177.3 | NP_080453.2 | 8 | 971 | ||
| GenBank mRNA | AK004707.1 | - | 8 | 951 | ||
| AK016681.1 | - | 8 | 978 | |||
| AK082912.1 | - | 8 | 952 | |||
| AK173238.1 | - | 8 | 916 | |||
| BC029731.1 | - | 8 | 932 | 4946215 | ||

Gpalpp1
Genomic Map