siRNA Details
| Gene Symbol: Abhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026179.2 | NP_080455.1 | 3 | 338 | ||
| GenBank mRNA | AK004873.1 | - | 3 | 224 | ||
| AK028453.1 | - | 3 | 224 | |||
| AK050377.1 | - | 3 | 223 | |||
| AK152717.1 | - | 4 | 272 | |||
| AK192646.1 | - | 1 | 37 | |||
| AK194347.1 | - | 3 | 152 | |||
| BC037063.1 | - | 3 | 220 | 4985992 | ||
| CN840045.1 | - | 3 | 349 | |||

Abhd5
Genomic Map