siRNA Details
| Gene Symbol: MARK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286128.1 | NP_001273057.1 | 4, 5 | 990 | ||
| GenBank mRNA | AB040910.1 | - | 4, 5 | 947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARK1| Gene Symbol: MARK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286128.1 | NP_001273057.1 | 4, 5 | 990 | ||
| GenBank mRNA | AB040910.1 | - | 4, 5 | 947 | ||