siRNA Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290338.1 | NP_001277267.1 | 2 | 327 | ||
| NM_001290339.1 | NP_001277268.1 | 2 | 282 | |||
| GenBank mRNA | AK012639.1 | - | 2 | 283 | ||
| AK089032.1 | - | 2 | 325 | |||
| BY771310.1 | - | 2 | 294 | |||

Zfand4
Genomic Map