siRNA Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290338.1 | NP_001277267.1 | 6 | 1196 | ||
| NM_001290339.1 | NP_001277268.1 | 4 | 896 | |||
| GenBank mRNA | AK012639.1 | - | 4 | 895 | ||
| AK089032.1 | - | 6 | 1194 | |||

Zfand4
Genomic Map