siRNA Details
| Gene Symbol: Tmem167b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026198.2 | NP_080474.2 | 2 | 239 | ||
| GenBank mRNA | AK008415.1 | - | 2 | 152 | ||
| AK046697.1 | - | 2 | 140 | |||
| BC050932.1 | - | 2 | 143 | 6811263 | ||
| BC055449.1 | - | 2 | 156 | 3993993 | ||
| BQ560740.1 | - | 2 | 239 | |||

Tmem167b
Genomic Map