siRNA Details
| Gene Symbol: Prl7c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026206.2 | NP_080482.1 | 5 | 575 | ||
| GenBank mRNA | AF525157.1 | - | 5 | 560 | ||
| AK005486.1 | - | 5 | 576 | |||
| AK014436.1 | - | 5 | 559 | |||
| BC099442.1 | - | 5 | 535 | 30716548 | ||

Prl7c1
Genomic Map