siRNA Details
| Gene Symbol: Saysd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026209.1 | NP_080485.1 | 1 | 242 | ||
| GenBank mRNA | AK007941.1 | - | 197 | |||
| AK015650.1 | - | 1 | 217 | |||
| AK031748.1 | - | 1 | 168 | |||
| BC027644.1 | - | 1 | 163 | 1248085 | ||

Saysd1
Genomic Map