siRNA Details
| Gene Symbol: Kctd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026214.3 | NP_080490.3 | 2 | 1005 | ||
| GenBank mRNA | AK008741.1 | - | 2 | 1000 | ||
| AK017592.1 | - | 1 | 736 | |||
| AK032314.1 | - | 1 | 712 | |||
| AK032624.1 | - | 1 | 743 | |||
| BC028748.1 | - | 1 | 720 | 1332429 | ||

Kctd4
Genomic Map