siRNA Details
Gene Symbol: Atg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026217.3 | NP_080493.2 | 2, 3 | 330 | ||
GenBank mRNA | AB066216.1 | - | 2, 3 | 302 | ||
AK005405.1 | - | 2, 3 | 334 | |||
AK008698.1 | - | 2, 3 | 306 | |||
AK016474.2 | - | 2, 3 | 309 | |||
AK167027.1 | - | 2, 3 | 308 | |||
AK198454.1 | - | 1, 2 | 60 | |||
AK208391.1 | - | 1, 2 | 60 | |||
AK214439.1 | - | 1, 2 | 71 | |||
BB853539.1 | - | 2, 3 | 331 | |||
BC070470.1 | - | 2, 3 | 300 | 30630196 |
