siRNA Details
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026229.4 | NP_080505.1 | 13, 14 | 1234 | ||
GenBank mRNA | AK002937.1 | - | 13, 14 | 1251 | ||
AK009734.1 | - | 12, 13 | 1114 | |||
AK016787.1 | - | 13, 14 | 1234 | |||
AK034931.1 | - | 900 | ||||
BC010729.1 | - | 13, 14 | 1227 | 3708143 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
