siRNA Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026231.2 | NP_080507.2 | 2 | 233 | ||
| GenBank mRNA | AK029724.1 | - | 2 | 232 | ||
| AK033409.1 | - | 2 | 271 | |||
| AK037169.1 | - | 2 | 272 | |||
| AK044441.1 | - | 2 | 241 | |||
| BC047136.1 | - | 2 | 233 | 6311866 | ||
| BC061240.1 | - | 2 | 229 | 6743615 | ||

Gstcd
Genomic Map