siRNA Details
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026231.2 | NP_080507.2 | 8 | 1623 | ||
GenBank mRNA | AK013967.1 | - | 5 | 475 | ||
AK017059.1 | - | 6 | 898 | |||
AK020021.1 | - | 7 | 1233 | |||
AK029724.1 | - | 8 | 1623 | |||
AK033409.1 | - | 8 | 1661 | |||
AK044441.1 | - | 8 | 1631 | |||
BC047136.1 | - | 8 | 1623 | 6311866 | ||
BC061240.1 | - | 8 | 1619 | 6743615 |
