siRNA Details
| Gene Symbol: MAT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005911.5 | NP_005902.1 | 5 | 843 | ||
| GenBank mRNA | AK291126.1 | - | 5 | 651 | ||
| AK293841.1 | - | 5 | 653 | |||
| AK297758.1 | - | 5 | 546 | |||
| AK308758.1 | - | 5 | 511 | |||
| AK309338.1 | - | 5 | 651 | |||
| AK316411.1 | - | 5 | 801 | |||
| BC001686.2 | - | 5 | 640 | 3010820 | ||
| BC001854.2 | - | 5 | 640 | 3010820 | ||
| BI549760.1 | - | 5 | 662 | |||
| DQ083239.1 | - | 5 | 579 | |||
| X68836.1 | - | 5 | 598 | |||

MAT2A
Genomic Map